Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzyloxy)benzenesulfonyl chloride 95%

Supplier: AMBEED, INC
Danger

A989385-1G A989385-250MG A989385-5G
77294-398EA 17 USD InStock
77294-398 77294-400 77294-402
4-(Benzyloxy)benzenesulfonyl chloride 95%
4-(Benzyloxy)benzenesulfonyl chloride
Precaución: for Research Use Only


Formula: C₁₃H₁₁ClO₃S
MW: 282.75 g/mol
MDL Number: MFCD04035557
CAS Number: 87001-32-9
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty