Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-0156 99%

Supplier: AMBEED, INC
Warning

A998216-10MG A998216-1MG A998216-25MG A998216-50MG A998216-5MG A998216-100MG A998216-250MG
77269-180EA 48 USD InStock
77269-180 77269-182 77269-184 77269-186 77269-188 77749-836 77749-837
AZD-0156 99%
AZD-0156
Precaución: for Research Use Only


Formula: C₂₆H₃₁N₅O₃
MW: 461.57 g/mol
MDL Number: MFCD30470661
CAS Number: 1821428-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty