Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(tert-Butyl)piperazine dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A998245-100MG A998245-1G A998245-250MG
77282-144EA 60 USD InStock
77282-144 77282-146 77282-148
2-(tert-Butyl)piperazine dihydrochloride 95%
2-(tert-Butyl)piperazine dihydrochloride
Precaución: for Research Use Only


Formula: C₈H₂₀Cl₂N₂
MW: 215.16 g/mol
MDL Number: MFCD16990497
CAS Number: 1159812-65-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty