Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Cytokeratin 8 + 18 + 19 Mouse Monoclonal Antibody [clone: ABT-CK18]

Supplier: ANTIBODIES.COM LLC
A95693-50 A95693-100
77051-322EA 356.8 USD InStock
77051-322 77051-320
Anti-Cytokeratin 8 + 18 + 19 Mouse Monoclonal Antibody [clone: ABT-CK18]
Antibodies
Mouse monoclonal (ABT-CK18) antibody to Cytokeratin 18 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: Cytokeratin 8 + 18 + 19
Clonality: Monoclonal
Clone: ABT-CK18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty