Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-GCDFP 15 Mouse Monoclonal Antibody [clone: ABT-GCDFP15]

Supplier: ANTIBODIES.COM LLC
A98368-50 A98368-100
77080-662EA 356.8 USD InStock
77080-662 77069-030
Anti-GCDFP 15 Mouse Monoclonal Antibody [clone: ABT-GCDFP15]
Antibodies
Mouse monoclonal (ABT-GCDFP15) antibody to GCDFP 15 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: GCDFP 15
Clonality: Monoclonal
Clone: ABT-GCDFP15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty