Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Complex IV Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A92884-100 A92884-50
77208-382EA 351.7 USD InStock
77208-382 77210-308
Anti-Complex IV Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Complex IV for WB and IHC with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500-1:1, 000, IHC: 1:100-1:200

Type: Primary
Antigen: Complex IV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty