Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-PCCB Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A92967-100 A92967-50
77205-406EA 494.4 USD InStock
77205-406 77209-780
Anti-PCCB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PCCB for WB and ICC/IF with samples derived from Human and Mouse.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:500-1:2, 000, ICC/IF: 1:50-1:200

Type: Primary
Antigen: PCCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty