Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-BAFF Rabbit Recombinant Antibody [clone: DM181]

Supplier: ANTIBODIES.COM LLC
A318528-100 A318528-500
77186-750EA 703.3 USD InStock
77186-750 77186-752
Anti-BAFF Rabbit Recombinant Antibody [clone: DM181]
Antibodies
Recombinant rabbit monoclonal [DM181] antibody to BAFF for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: BAFF
Clonality: Recombinant
Clone: DM181
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty