Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Phospholipase C beta 2/PLCB2 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A10313-100
76849-160EA 351.7 USD InStock
76849-160 76849-626
Anti-Phospholipase C beta 2/PLCB2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Phospholipase C beta 2 / PLCB2 for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:1000

Type: Primary
Antigen: Phospholipase C beta 2/PLCB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty