Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IKK alpha Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A10986-100
76857-744EA 458.7 USD InStock
76857-744 76857-226
Anti-IKK alpha Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IKK alpha (phospho Ser180) for WB with samples derived from Human and Mouse.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: IKK alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty