Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-NDUFS5 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A307601-100 A307601-50
77226-636EA 351.7 USD InStock
77226-636 77226-638
Anti-NDUFS5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to NDUFS5 for WB and ICC/IF with samples derived from Human.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:500 to 1:1,000, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: NDUFS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty