Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Alcohol Dehydrogenase Rabbit Monoclonal Antibody [clone: ARC2203]

Supplier: ANTIBODIES.COM LLC
A307868-100 A307868-50
77227-684EA 494.4 USD InStock
77227-684 77227-686
Anti-Alcohol Dehydrogenase Rabbit Monoclonal Antibody [clone: ARC2203]
Antibodies
Rabbit monoclonal [ARC2203] antibody to Alcohol Dehydrogenase for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2,000

Type: Primary
Antigen: Alcohol Dehydrogenase
Clonality: Monoclonal
Clone: ARC2203
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty