Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-beta III Tubulin Mouse Monoclonal Antibody [clone: AMC0106]

Supplier: VWR
77230-616EA 470 USD InStock
77230-616 77230-618
Anti-beta III Tubulin Mouse Monoclonal Antibody [clone: AMC0106]
Antibodies
Mouse monoclonal [AMC0106] antibody to beta III Tubulin for WB and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:1,000 to 1:5,000, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: beta III Tubulin
Clonality: Monoclonal
Clone: AMC0106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty