Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Insulin Receptor alpha + IGF1 REceptor Rabbit Monoclonal Antibody [clone: ARC54074]

Supplier: ANTIBODIES.COM LLC
A309683-100
77215-240EA 494.4 USD InStock
77215-240 77210-390
Anti-Insulin Receptor alpha + IGF1 REceptor Rabbit Monoclonal Antibody [clone: ARC54074]
Antibodies
Rabbit monoclonal [ARC54074] antibody to Insulin Receptor alpha + IGF1 REceptor for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:1,000

Type: Primary
Antigen: Insulin Receptor alpha + IGF1 REceptor
Clonality: Monoclonal
Clone: ARC54074
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty