Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-AMPK alpha 1 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A309813-100
77204-182EA 351.7 USD InStock
77204-182 77205-578
Anti-AMPK alpha 1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to AMPK alpha 1 for WB with samples derived from Mouse.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:100 to 1:500

Type: Primary
Antigen: AMPK alpha 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty