Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IL-1 beta Rabbit Monoclonal Antibody [clone: ARC56185]

Supplier: VWR
77209-876EA 470 USD InStock
77209-876 77212-832
Anti-IL-1 beta Rabbit Monoclonal Antibody [clone: ARC56185]
Antibodies
Rabbit monoclonal [ARC56185] antibody to IL-1 beta for WB with samples derived from Mouse.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:1,000 to 1:5,000

Type: Primary
Antigen: IL-1 beta
Clonality: Monoclonal
Clone: ARC56185
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty