Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD137 Rabbit Recombinant Antibody [clone: DM66]

Supplier: ANTIBODIES.COM LLC
A317400-500 A317400-100
77182-236EA 1967.3 USD InStock
77182-236 77182-234
Anti-CD137 Rabbit Recombinant Antibody [clone: DM66]
Antibodies
Recombinant rabbit monoclonal [DM66] antibody to CD137 for ELISA and Flow Cytometry with samples derived from Human.

  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100

Type: Primary
Antigen: CD137
Clonality: Recombinant
Clone: DM66
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty