Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-beta glucuronidase (GUSB) Rabbit Monoclonal Antibody [clone: ARC2408]

Supplier: ANTIBODIES.COM LLC
A306996-100
77224-244EA 494.4 USD InStock
77224-244 77224-246
Anti-beta glucuronidase (GUSB) Rabbit Monoclonal Antibody [clone: ARC2408]
Antibodies
Rabbit monoclonal [ARC2408] antibody to beta glucuronidase (GUSB) for WB with samples derived from Human.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:1,000

Type: Primary
Antigen: beta glucuronidase (GUSB)
Clonality: Monoclonal
Clone: ARC2408
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty