Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-COX IV Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A9916-100 A9916-50
76866-690EA 351.7 USD InStock
76866-690 76868-974
Anti-COX IV Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to COX IV for WB, ICC/IF and IP with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ICC/IF, IP
  • Recommended Dilutions: WB: 1:1000 to 1:5000, ICC/IF: 1:50 to 1:200, IP: 1:500 to 1:100

Type: Primary
Antigen: COX IV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty