Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Ran Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A12734-100
76853-422EA 351.7 USD InStock
76853-422 76857-182
Anti-Ran Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Ran for WB and ICC/IF with samples derived from Human, Mouse, Rat and Monkey.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:100 to 1:500, ICC/IF: 1:50 to 1:20

Type: Primary
Antigen: Ran
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty