Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgG H&L Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: ANTIBODIES.COM LLC
A17346-100 A17346-200
77217-278EA 107 USD InStock
77217-278 77217-280
Anti-IgG H&L Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Goat Anti-Rabbit IgG H&L Antibody (TRITC) for IHC, ICC/IF and Flow Cytometry with samples derived from Rabbit.

  • Validated Applications: IHC, ICC/IF, Flow Cytometry
  • Recommended Dilutions: IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200, Flow Cytometry: 1:50 to 1:20

Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty