Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-beta Tubulin Mouse Monoclonal Antibody [clone: 1B12]

Supplier: ANTIBODIES.COM LLC
A85428-50 A85428-100
77065-738EA 428.1 USD InStock
77065-738 77078-322
Anti-beta Tubulin Mouse Monoclonal Antibody [clone: 1B12]
Antibodies
Mouse monoclonal (1B12) antibody to beta Tubulin for WB, ICC/IF and IHC with samples derived from Human, Monkey, Rat and Mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:5000 to 1:10000, ICC/IF: 1:500

Type: Primary
Antigen: beta Tubulin
Clonality: Monoclonal
Clone: 1B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Monkey;Rat;Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty