Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromo-4-(4,4-dimethylcyclohexyl)phenol ≥97%

Supplier: AOB CHEM USA
Warning

109972-500MG 109972-1G 109972-25G 109972-5G
77323-466EA 293.55 USD InStock
77323-466 77347-534 77383-478 77371-542
2,6-Dibromo-4-(4,4-dimethylcyclohexyl)phenol ≥97%
2,6-Dibromo-4-(4,4-dimethylcyclohexyl)phenol
Precaución: for Research Use Only


Formula: C₁₄H₁₈Br₂O
Storage Temperature: Ambient
MDL Number: MFCD34780376
CAS Number: 2901099-39-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty