Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Fluoro-6-(trifluoromethyl)phenyl)ethanol ≥97%

Supplier: AOB CHEM USA
Warning

114028-500MG 114028-1G 114028-5G 114028-25G
77323-536EA 276.04 USD InStock
77323-536 77347-618 77359-646 77383-518
1-(2-Fluoro-6-(trifluoromethyl)phenyl)ethanol ≥97%
1-(2-Fluoro-6-(trifluoromethyl)phenyl)ethanol
Precaución: for Research Use Only


Formula: C₉H₈F₄O
Storage Temperature: Ambient
MDL Number: MFCD34148935
CAS Number: 1503305-18-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty