Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(5-Bromo-2-fluorophenyl)-1,1,1-trifluoropropan-2-ol ≥97%

Supplier: AOB CHEM USA
Warning

116692-500MG 116692-1G 116692-5G 116692-25G
77323-578EA 244.11 USD InStock
77323-578 77347-658 77359-700 77383-546
2-(5-Bromo-2-fluorophenyl)-1,1,1-trifluoropropan-2-ol ≥97%
2-(5-Bromo-2-fluorophenyl)-1,1,1-trifluoropropan-2-ol
Precaución: for Research Use Only


Formula: C₉H₇BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD24341881
CAS Number: 1553415-03-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty