Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-(3-bromophenyl)-3-hydroxybutanoate ≥97%

Supplier: AOB CHEM USA
Warning

120614-500MG 120614-1G 120614-5G 120614-25G
77323-692EA 99.91 USD InStock
77323-692 77342-908 77359-840 77371-828
Methyl 3-(3-bromophenyl)-3-hydroxybutanoate ≥97%
Methyl 3-(3-bromophenyl)-3-hydroxybutanoate
Precaución: for Research Use Only


Formula: C₁₁H₁₃BrO₃
Storage Temperature: Ambient
CAS Number: 2918953-29-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty