Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoronicotinonitrile

Supplier: AOB CHEM USA
Warning

137579-500MG 137579-1G 137579-5G 137579-25G
77323-902EA 118.45 USD InStock
77323-902 77343-540 77360-092 77372-058
2,6-Difluoronicotinonitrile
2,6-Difluoronicotinonitrile
Precaución: for Research Use Only


Formula: C₆H₂F₂N₂
Storage Temperature: Ambient
MDL Number: MFCD13185485
CAS Number: 869557-17-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty