Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyclopropylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

164675-500MG 164675-1G 164675-5G 164675-25G
77324-078EA 122.57 USD InStock
77324-078 77344-020 77360-316 77372-272
2-Cyclopropylbenzoic acid ≥97%
2-Cyclopropylbenzoic acid


Formula: C₁₀H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD01550965
CAS Number: 3158-74-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty