Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine, 3-chloro-2,6-difluoro-4-iodo- ≥97%

Supplier: AOB CHEM USA
Warning

18658-500MG 18658-1G 18658-5G 18658-25G
77324-184EA 165.83 USD InStock
77324-184 77346-160 77360-456 77383-958
Pyridine, 3-chloro-2,6-difluoro-4-iodo- ≥97%
Pyridine, 3-chloro-2,6-difluoro-4-iodo-
Precaución: for Research Use Only


Formula: C₅HClF₂IN
MW: 275.422 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756674
CAS Number: 685517-73-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty