Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4'-Isobutoxyphenyl)phenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

21966-500MG 21966-1G 21966-5G 21966-25G
77324-356EA 222.48 USD InStock
77324-356 77348-250 77360-676 77384-090
4-(4'-Isobutoxyphenyl)phenylboronic acid ≥97%
4-(4'-Isobutoxyphenyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD18383567
CAS Number: 1235568-92-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty