Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-isopropoxynicotinic acid ≥97%

Supplier: AOB CHEM USA
Warning

23377-500MG 23377-1G 23377-5G 23377-25G
77324-434EA 154.5 USD InStock
77324-434 77345-538 77360-766 77384-150
2-Fluoro-5-isopropoxynicotinic acid ≥97%
2-Fluoro-5-isopropoxynicotinic acid
Precaución: for Research Use Only


Formula: C₉H₁₀FNO₃
MW: 199.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056756
CAS Number: 1370025-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty