Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-3-chlorobenzoic acid methyl ester ≥97%

Supplier: AOB CHEM USA
Warning

23728-500MG 23728-1G 23728-5G 23728-25G
77324-476EA 236.9 USD InStock
77324-476 77348-346 77360-802 77384-178
4-Benzyloxy-3-chlorobenzoic acid methyl ester ≥97%
4-Benzyloxy-3-chlorobenzoic acid methyl ester
Precaución: for Research Use Only


Formula: C₁₅H₁₃ClO₃
Storage Temperature: Ambient
MDL Number: MFCD06203772
CAS Number: 536974-86-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty