Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium trifluoro(6-fluoro-5-methylpyridin-3-yl)borate ≥97%

Supplier: AOB CHEM USA
Warning

24022-1G 24022-5G 24022-25G
77341-908EA 108.15 USD InStock
77341-908 77360-840 77372-660
Potassium trifluoro(6-fluoro-5-methylpyridin-3-yl)borate ≥97%
Potassium trifluoro(6-fluoro-5-methylpyridin-3-yl)borate
Precaución: for Research Use Only


Formula: C₆H₅BF₄KN+
Storage Temperature: Ambient
MDL Number: MFCD09993153
CAS Number: 1262538-12-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty