Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium trifluoro(6-methoxynaphthalen-2-yl)boranuide ≥95%

Supplier: AOB CHEM USA
Warning

24703-500MG 24703-1G 24703-5G
77324-572EA 319.3 USD InStock
77324-572 77348-424 77372-706
Potassium trifluoro(6-methoxynaphthalen-2-yl)boranuide ≥95%
Potassium trifluoro(6-methoxynaphthalen-2-yl)boranuide
Precaución: for Research Use Only


Formula: C₁₁H₉BF₃KO
Storage Temperature: Ambient
MDL Number: MFCD19514692
CAS Number: 1163708-25-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty