Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl[(3-Bromo-4-methylphenyl)methyl]amine ≥97%

Supplier: AOB CHEM USA
Warning

24954-500MG 24954-1G 24954-5G 24954-25G
77324-610EA 199.82 USD InStock
77324-610 77348-456 77360-940 77384-294
Diethyl[(3-Bromo-4-methylphenyl)methyl]amine ≥97%
Diethyl[(3-Bromo-4-methylphenyl)methyl]amine
Precaución: for Research Use Only


Formula: C₁₂H₁₈BrN
MW: 256.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628244
CAS Number: 1414870-79-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty