Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3-isopropylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

25825-500MG 25825-1G 25825-5G 25825-25G
77324-726EA 210.12 USD InStock
77324-726 77348-578 77361-046 77384-378
4-Hydroxy-3-isopropylphenylboronic acid ≥97%
4-Hydroxy-3-isopropylphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₃BO₃
MW: 180.009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414526
CAS Number: 1451390-86-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty