Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-(methylthio)-benzoic acid methyl ester ≥95%

Supplier: AOB CHEM USA
Warning

26018-500MG 26018-1G 26018-5G
77335-532EA 357.41 USD InStock
77335-532 77348-590 77372-838
3-Chloro-4-(methylthio)-benzoic acid methyl ester ≥95%
3-Chloro-4-(methylthio)-benzoic acid methyl ester
Precaución: for Research Use Only


Formula: C₉H₉ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD28977205
CAS Number: 105442-23-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty