Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-chloro-5-methylphenyboronic acid, pinacol ester ≥95%

Supplier: AOB CHEM USA
Warning

26234-500MG 26234-1G 26234-5G
77324-780EA 334.75 USD InStock
77324-780 77348-624 77372-864
2-Amino-3-chloro-5-methylphenyboronic acid, pinacol ester ≥95%
2-Amino-3-chloro-5-methylphenyboronic acid, pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₉BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD24039662
CAS Number: 2121513-87-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty