Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-methoxy-α-methylbenzyl alcohol ≥95%

Supplier: AOB CHEM USA
Warning

28162-500MG 28162-1G 28162-5G 28162-25G
77324-926EA 91.67 USD InStock
77324-926 77342-418 77361-250 77373-008
2-Fluoro-4-methoxy-α-methylbenzyl alcohol ≥95%
2-Fluoro-4-methoxy-α-methylbenzyl alcohol
Precaución: for Research Use Only


Formula: C₉H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD12047707
CAS Number: 74457-87-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty