Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1H-imidazole-2-carbaldehyde oxime ≥97%

Supplier: AOB CHEM USA
Warning

28421-500MG 28421-1G 28421-5G 28421-25G
77324-944EA 238.96 USD InStock
77324-944 77348-770 77361-266 77384-520
1-Methyl-1H-imidazole-2-carbaldehyde oxime ≥97%
1-Methyl-1H-imidazole-2-carbaldehyde oxime
Precaución: for Research Use Only


Formula: C₅H₇N₃O
MW: 125.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21336097
CAS Number: 20062-62-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty