Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyclobutylphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

28881-500MG 28881-1G 28881-5G 28881-25G
77324-974EA 247.2 USD InStock
77324-974 77348-778 77361-306 77384-540
3-Cyclobutylphenylboronic acid pinacol ester ≥97%
3-Cyclobutylphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₆H₂₃BO₂
Storage Temperature: Ambient
MDL Number: MFCD18733633
CAS Number: 2121514-70-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty