Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxyphenoxyacetaldehyde diethyl acetal ≥97%

Supplier: AOB CHEM USA
Warning

29608-500MG 29608-1G 29608-5G 29608-25G
77325-034EA 151.41 USD InStock
77325-034 77345-418 77361-366 77384-582
4-Methoxyphenoxyacetaldehyde diethyl acetal ≥97%
4-Methoxyphenoxyacetaldehyde diethyl acetal
Precaución: for Research Use Only


Formula: C₁₃H₂₀O₄
MW: 240.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192269
CAS Number: 69034-13-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty