Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-Methyl-2-propan-2-yloxyphenyl)methanol ≥97%

Supplier: AOB CHEM USA
Warning

29659-500MG 29659-1G 29659-5G 29659-25G
77325-044EA 149.35 USD InStock
77325-044 77345-294 77361-376 77384-592
(5-Methyl-2-propan-2-yloxyphenyl)methanol ≥97%
(5-Methyl-2-propan-2-yloxyphenyl)methanol
Precaución: for Research Use Only


Formula: C₁₁H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD18331439
CAS Number: 1343277-94-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty