Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-(methylsulfanyl)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

30394-500MG 30394-1G 30394-5G 30394-25G
77325-104EA 156.56 USD InStock
77325-104 77345-602 77361-436 77384-634
2-Fluoro-6-(methylsulfanyl)benzoic acid ≥97%
2-Fluoro-6-(methylsulfanyl)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₇FO₂S
MW: 186.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21944950
CAS Number: 1516052-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty