Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isopropoxy-3-pyridineboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

30531-1G 30531-5G 30531-25G
77341-860EA 106.09 USD InStock
77341-860 77361-448 77373-162
2-Isopropoxy-3-pyridineboronic acid pinacol ester ≥97%
2-Isopropoxy-3-pyridineboronic acid pinacol ester


Formula: C₁₄H₂₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD07366722
CAS Number: 848243-25-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty