Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-nicotinic acid methyl ester ≥97%

Supplier: AOB CHEM USA
Warning

30537-500MG 30537-1G 30537-5G 30537-25G
77325-112EA 160.68 USD InStock
77325-112 77345-762 77361-450 77384-642
2-Benzyloxy-nicotinic acid methyl ester ≥97%
2-Benzyloxy-nicotinic acid methyl ester
Precaución: for Research Use Only


Formula: C₁₄H₁₃NO₃
MW: 243.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088162
CAS Number: 52513-17-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty