Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-3-chloro-2,4-difluorobenzyl alcohol ≥95%

Supplier: AOB CHEM USA
Warning

30819-500MG 30819-1G 30819-5G 30819-25G
77325-158EA 208.06 USD InStock
77325-158 77348-928 77361-494 77384-676
6-Bromo-3-chloro-2,4-difluorobenzyl alcohol ≥95%
6-Bromo-3-chloro-2,4-difluorobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₄BrClF₂O
MW: 257.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723163
CAS Number: 2121512-37-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty