Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-Bromo-6-fluoro-3-(methylsulfanyl)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

30958-500MG 30958-1G 30958-5G 30958-25G
77325-188EA 181.28 USD InStock
77325-188 77346-800 77361-526 77384-700
Methyl 2-Bromo-6-fluoro-3-(methylsulfanyl)benzoate ≥97%
Methyl 2-Bromo-6-fluoro-3-(methylsulfanyl)benzoate
Precaución: for Research Use Only


Formula: C₉H₈BrFO₂S
Storage Temperature: Ambient
MDL Number: MFCD29088239
CAS Number: 1879026-08-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty