Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-6-isopropoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

31364-500MG 31364-1G 31364-5G 31364-25G
77325-250EA 181.28 USD InStock
77325-250 77346-804 77361-606 77384-744
2,3-Difluoro-6-isopropoxybenzoic acid ≥97%
2,3-Difluoro-6-isopropoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD29915588
CAS Number: 1917346-66-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty