Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethyl-4-methoxyphenylboronic acid piancol ester ≥97%

Supplier: AOB CHEM USA
Warning

31762-500MG 31762-1G 31762-25G 31762-5G
77325-304EA 285.31 USD InStock
77325-304 77349-038 77384-790 77373-296
3-Ethyl-4-methoxyphenylboronic acid piancol ester ≥97%
3-Ethyl-4-methoxyphenylboronic acid piancol ester
Precaución: for Research Use Only


Formula: C₁₅H₂₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD22414611
CAS Number: 2121511-72-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty